Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RHPS 4 methosulfate
GREENER_CHOICESDP

Catalog No. 531150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-115-0 50 mg
53-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-115-0 Supplier Tocris Bioscience™ Supplier No. 5311/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Telomerase inhibitor

RHPS 4 methosulfate is a telomerase inhibitor. Inhibits growth of medulloblastoma and glioblastoma cells in vitro. Induces telomere injury and apoptosis, and reduces growth in CG5 breast cancer cell xenografts in mice. Sensitizes radiation resistant glioblastoma cell lines to radiation.

Specifications

Chemical Name or Material 3,11-Difluoro-6,8,13-trimethylquino[4,3,2-kl]acridinium methylsulfate
CAS 390362-78-4
Quantity 50 mg
Target Telomerase Inhibitors
Molecular Formula C22H17F2N2.CH3O4S
Formula Weight 458.48
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.