Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human uPAR His-tag Avi-tag Protein
Click to view available options
Quantity:
100 μg
Description
- uPA is synthesized and released from cells as a single-chain (sc) pro-enzyme with limited enzymatic activity and is converted to an active two-chain (tc) disulfide-linked active enzyme by plasmin and other specific proteinases.
- Both the scuPA and tcuPA bind with high-affinity to the cell surface via the glycosyl phosphatidylinositol-linked receptor uPAR which serves to localize the uPA proteolytic activity.
- The enzymatic activity of scuPA has also been shown to be enhanced by binding to uPAR.
- Independent of their proteolytic activity, the uPA/uPAR interaction also initiates signal transduction responses resulting in activation of protein tyrosine kinases, gene expression, cell adhesion, and chemotaxis.
- uPAR can interact with integrins to suppress normal integrin adhesive function and promote adhesion to vitronectin through a high affinity vitronectin binding site on uPAR.
- uPAR cDNA encodes a 335 amino acid (aa) residue precursor protein with a 22 aa residue signal peptide, five potential N-linked glycosylation sites and a C-terminal GPI-anchor site.
- An alternate spliced variant of uPAR encoding a secreted soluble form of uPAR also exists.
- Human and mouse uPAR share approximately 60% aa sequence identity and the receptor-ligand interaction is strictly species-specific.
Specifications
Specifications
Accession Number | Q03405.1 |
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 5329 |
Molecular Weight (g/mol) | 29 kDa |
Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
Quantity | 100 μg |
Source | Human embryonic kidney cell, HEK293-derived human uPAR protein, (N-terminus) Human uPAR (Leu23-Arg303) Accession # Q03405.1 HHHHHH Avi-tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
Conjugate | Biotin |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction