Description
- For use in density comparison with other igneous rocks
- Correlates well to Next Generation Science Standards Evidence Statements
- High silica content makes it difficult for this rock to erupt, leading to explosive eruptions that emit ash and dust into atmosphere
- Evolved chemistry makes it lightweight relative to other igneous rocks, good identifier during classification exercises
Specifications
Specifications
| Product Type | Igneous Rock |
| For Use With (Application) | Density comparison with other igneous rocks |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 1 Ea. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Rhyolite |