Learn More
Mopec™ Rhythm Manual Microtome
SDP
Catalog No. 22445678 Shop All Mopec Products
Click to view available options
Profile:
Low
Catalog No. 22-445-678
Supplier: Mopec™ BK125
Only null left
Add to Cart
Add to Cart
Built specifically for use in histology and pathology laboratories, the Rhythm Microtome features a manual operation system and durable construction, giving the user precise control for the sectioning process.
Designed with reliability in mind, this simple-to-use microtome is built to be more efficient and consistent in microtomy.
  • Consistent block alignment
  • Quick cassette clamp
  • Enhanced user control
  • Ergonomic hand wheel
  • Longer blade life 2-in-1 blade holder
  • Clean sectioning
  • Manual coarse feed
  • Waste tray
  • ±1% precision error
  • XY 8° specimen orientation
  • Lateral movement of blade holder: 0 to 23 mm
Specifications
Profile Low
For Use With (Equipment) Histology and Pathology Equipment
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.