Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIAM/APBB1IP Antibody (7E7A3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252417
Description
RIAM/APBB1IP Monoclonal specifically detects RIAM/APBB1IP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
RIAM/APBB1IP | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
amyloid beta (A4) precursor protein-binding, family B, member 1 interactingprotein, amyloid beta A4 precursor protein-binding family B member 1-interacting protein, APBB1-interacting protein 1, INAG1, PREL1, PREL-1, proline rich EVH1 ligand 1, Proline-rich EVH1 ligand 1, Proline-rich protein 73, Rap1-GTP-interacting adapter molecule, Rap1-interacting adaptor molecule, RARP1, Retinoic acid-responsive proline-rich protein 1, RIAMRARP-1 | |
Mouse | |
73.2 kDa | |
0.1 mg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
7E7A3 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
APBB1IP | |
Purified recombinant fragment of human APBB1IP (AA: 1-151) expressed in E. Coli. | |
Protein A or G purified | |
RUO | |
54518 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction