Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIAM/APBB1IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP21430025UL
Description
RIAM/APBB1IP Polyclonal specifically detects RIAM/APBB1IP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RIAM/APBB1IP | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
amyloid beta (A4) precursor protein-binding, family B, member 1 interactingprotein, amyloid beta A4 precursor protein-binding family B member 1-interacting protein, APBB1-interacting protein 1, INAG1, PREL1, PREL-1, proline rich EVH1 ligand 1, Proline-rich EVH1 ligand 1, Proline-rich protein 73, Rap1-GTP-interacting adapter molecule, Rap1-interacting adaptor molecule, RARP1, Retinoic acid-responsive proline-rich protein 1, RIAMRARP-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
54518 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
APBB1IP | |
This antibody was developed against a recombinant protein corresponding to the amino acids: MGESSEDIDQMFSTLLGEMDLLTQSLGVDTLPPPDPNPPRAEFNYSVGFKDLNESLNALEDQDLDALMADLVADISEAEQRTIQAQKESLQNQHHSASLQASIFSGAASLGYGTNVAATGISQYEDDLPPPPADPVLDLPLPPP | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction