Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547911
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547911 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547911 Supplier Invitrogen™ Supplier No. PA547911
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This antibody detects recombinant human RIAM/APBB1IP in direct ELISAs. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIAM
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene APBB1IP
Gene Accession No. Q7Z5R6, Q8R5A3
Gene Alias 9930118P07Rik; amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein; amyloid beta A4 precursor protein-binding family B member 1-interacting protein; amyloid beta precursor protein binding family B member 1 interacting protein; APBB1-interacting protein 1; Apbb1ip; INAG1; Prel1; PREL-1; proline rich EVH1 ligand 1; proline-rich EVH1 ligand 1; Proline-rich protein 48; proline-rich protein 73; Prp48; Rap1-GTP-interacting adapter molecule; Rap1-GTP-interacting adaptor molecule; Rap1-interacting adaptor molecule; RARP1; RARP-1; retinoic acid-responsive proline-rich protein 1; RIAM; RIAMRARP-1; zgc:63968
Gene Symbols APBB1IP
Host Species Sheep
Immunogen E. coli-derived recombinant human RIAM/APBB1IP Gly2-Ser89.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54518, 54519
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.