Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribavirin, MP Biomedicals™

Catalog No. MP219606650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219606650 50 mg
MP219606610 10 mg
MP219606683 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219606650 Supplier MP Biomedicals, Inc Supplier No. 0219606650
Add to Cart
Add to Cart

Active against many DNA and RNA viruses. Ribavirin, MP Biomedicals is an antiviral compound that is active against RSV, HIV, HCV and other infectious agents. It is a nucleoside antimetabolite drug that interferes with duplication of viral genetic material

Ribavirin is used as an antiviral agent against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. It inhibits purine (GTP pools) biosynthesis and is a precursor to the di- and tri-phosphates that inhibit viral RNA-dependent RNA polymerases. Its metabolite, ribavirin 5'-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase, but many other mechanisms of action are also supported with experimental evidence. It is also an analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.207
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
Synonym ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
PubChem CID 37542
ChEBI CHEBI:63580
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N

Specifications

Melting Point 173°C (literature)
Loss on Drying 0.04% (Lit.)
Quantity 50 mg
Solubility Information Soluble in water (20mg/mL, clear, colorless solution).
Formula Weight 244.2g/mol
Percent Purity >99%
Physical Form Powder
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.