Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribavirin, USP, 98.9-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18606076 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

R1052, 36791-04-5, C8H12N4O5

Spectrum™ Chemical Ribavirin, USP is an antiviral drug used primarily to treat hepatitis C and viral hemorrhagic fevers. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.21
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide
SMILES NC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

CAS Max % 1
Ignition Residue 0.0025
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 1 kg
Specific Rotation -33.5° to -37.0°
Percent Purity 98.9 to 101.5%
Grade USP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.