Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIBC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564876
Description
Immunogen sequence: CERQREQKANL AEIQHQSTSD LLTENPQVAQ HPMAPYRVLP YCWKGMTPEQ QAAIRKEQEV QRSKKQAHRQ AEKTLDTEWK SQTMSSAQAV LELEEQEREL CAVFQRGLGS FNQ Highest antigen sequence identity to the following orthologs - mouse 67%, rat 70%.
RIBC1 is a protein coding gene.
Specifications
RIBC1 | |
Polyclonal | |
Unconjugated | |
RIBC1 | |
2610028I09Rik; RIB43A domain with coiled-coils 1; RIB43A-like with coiled-coils protein 1; RIBC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
158787 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N443 | |
RIBC1 | |
Recombinant Human RIBC1. Recombinant protein control fragment (Product #RP-105681). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction