Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIBC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551861 Supplier Invitrogen™ Supplier No. PA551861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKIMCILENR KKRDRKNLCR AINDFQQSFQ KPETRREFDL SDPLALKKDL PARQSDNDVR NTISGMQKFM GEDLNFHERK KFQEEQNREW SLQQQREWKN ARAEQKCAEA LYTETRLQFD ETAKHLQKLE STTRKAVCAS VKDF Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 76%.

RIBC2 (RIB43A Domain With Coiled-Coils 2) is a Protein Coding gene. An important paralog of this gene is RIBC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIBC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RIBC2
Gene Accession No. Q9H4K1
Gene Alias 4930579A10Rik; C22orf11; RGD1309033; RIB43A domain with coiled-coils 2; RIB43A-like with coiled-coils protein 2; RIBC2; testis-specific ribbon protein; Trib
Gene Symbols RIBC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIBC2. Recombinant protein control fragment (Product #RP-91515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.