Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIBC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598988
Description
RIBC2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
RIBC2 (RIB43A Domain With Coiled-Coils 2) is a Protein Coding gene. An important paralog of this gene is RIBC1.
Specifications
| RIBC2 | |
| Polyclonal | |
| Unconjugated | |
| RIBC2 | |
| 4930579A10Rik; C22orf11; RGD1309033; RIB43A domain with coiled-coils 2; RIB43A-like with coiled-coils protein 2; RIBC2; testis-specific ribbon protein; Trib | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26150 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H4K1 | |
| RIBC2 | |
| Recombinant Human RIB43A-like with coiled-coils protein 2 protein (1-309AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction