Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ribH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144589
Description
ribH Polyclonal Antibody for Western Blot, ELISA
Catalyzes the formation of 6,7-dimethyl-8-ribityllumazine by condensation of 5-amino-6-(D-ribitylamino)uracil with 3,4-dihydroxy-2-butanone 4-phosphate. This is the penultimate step in the biosynthesis of riboflavin.
Specifications
ribH | |
Polyclonal | |
Unconjugated | |
ribH | |
6,7-dimethyl-8-ribityllumazine synthase; ribH | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1192672 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
O66529 | |
ribH | |
Recombinant Aquifex aeolicus ribH protein (1-154aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction