Learn More
Riboflavin, 98%
Description
Photoinitiator of polymerization for PAGE gelsRiboflavin is used in the preparation of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). It also provides protection against oxidative damage. It is also used as a food coloring and a vitamin. Further, it is employed as a medication, animal feed supplement and nutritional supplement. In addition to this, it is used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water, absolute ethanol and dimethyl sulfoxide. Insoluble in acetone and chloroform.
Notes
Store in a cool place. Incompatible with Strong oxidizing agents, reducing agents, bases, calcium and metallic salts.
Specifications
Specifications
| Melting Point | 290°C |
| Assay Percent Range | 98% |
| Quantity | 100 g |
| Sensitivity | Light sensitive |
| Merck Index | 14,8200 |
| Solubility Information | Soluble in water,absolute ethanol and dimethyl sulfoxide. Insoluble in acetone and chloroform. |
| Formula Weight | 376.37 |
| Percent Purity | 98% |
| Chemical Name or Material | Riboflavin |
Safety and Handling
| EINECSNumber | 201-507-1 |
| RTECSNumber | VJ1400000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.