Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribonuclease A Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300131UV
Description
Ribonuclease A Polyclonal specifically detects Ribonuclease A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ribonuclease A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 | |
Produced in rabbits immunized with purified, recombinant Ribonuclease A (P07998; Met1-Thr156). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
6035 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction