Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Ribonuclease A (Bovine)

Catalog No. 89960020
Click to view available options
:
25mg

Used for Enzyme

#REF!

Specifications

Accession Number P61823
For Use With (Application) Enzyme Activity
Formulation In 100mM phosphate buffer containing 0.1% v/v phenol as a preservative
Gene ID (Entrez) 282340
Molecular Weight (g/mol) 13.7kDa
Name Ribonuclease A (Bovine)
Preparation Method Native protein chromatographically purified from Bovine Pancreas
Quantity 25 mg
Storage Requirements Store at -20°C.
Regulatory Status RUO
Gene Alias RNS1
Common Name RNASE1
Gene Symbol RNASE1
Assays Ribonuclease Assay; RNA removal during DNA isolation; RNA sequence analysis; RNase protection assays; RNA quantification or mapping.
Biological Activity ≥3,000 units per mg protein. One Kunitz unit is the amount of enzyme required to cause an increase in absorbance of 1.0 at 260 nm at 37°C (pH 5.0) when yeast rRNA is hydrolyzed to acid-soluble oligonucleotides. 50 units are equivalent to 1 Kunitz unit.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Bovine Pancreas
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.