Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribonuclease A Polyclonal Antibody, Biotin, Rockland™
SDP

Catalog No. 501991211
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1211 25 μL
50-199-1210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1211 Supplier Rockland Immunochemicals Supplier No. 2004688S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Ribonuclease A antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Rabbit Serum as well as purified and partially purified Ribonuclease A [Bovine Pancreas]. Cross reactivity against Ribonuclease A from other sources may occur but have not been specifically determined.

Endonuclease that catalyzes the cleavage of RNA on the 3' side of pyrimidine nucleotides. Acts on single stranded and double stranded RNA.

Specifications

Antigen Ribonuclease A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Gene RNASE1
Gene Accession No. P61823
Gene Alias AI574248; BOS_10503; BOS_10505; HP-RNase; RAC1; Rib 1; RIB1; RIB-1; Ribonuclease 1; ribonuclease 1, pancreatic; Ribonuclease A; ribonuclease A (pancreatic); ribonuclease A C1; ribonuclease A family member 1, pancreatic; Ribonuclease BS-1; Ribonuclease pancreatic; ribonuclease, RNase A family, 1 (pancreatic); RNase 1; RNase A; RNase UpI-1; RNASE1; RNAseA; RNS1; Seminal ribonuclease; seminal RNase; SRN; S-RNase
Gene Symbols RNASE1
Host Species Rabbit
Immunogen Ribonuclease A [Bovine Pancreas]
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280930, 282340
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.