Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Ribonuclease H

Catalog No. PRM4285 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 U
50 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-M4285 250 U
PR-M4281 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-M4285 Supplier Promega Supplier No. M4285
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNase H is an endonuclease that specifically hydrolyzes the phosphodiester bonds of RNA hybridized to DNA.

  • Endonuclease that Specifically Hydrolyzes the Phosphodiester Bonds of RNA Hybridized to DNA
  • Removes the RNA strand prior to second-strand cDNA synthesis
  • Analyzes in vitro polyadenylation reaction products
  • Supplied at a concentration of 0.5-2u/μL

Specifications

Concentration 0.5 to 2 U/μL
Color Colorless
pH 7.8
For Use With (Application) Removal of the RNA strand prior to second strand cDNA synthesis, analysis of in-vitro polyadenylation reaction products, microarrays
Storage Buffer 20mM HEPES-KOH (pH 7.8 at 25°C), 1mM DTT, 50mM KCl, 0.2mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 250 U
Product Type Ribonuclease H
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.