Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribonuclease Inhibitor Antibody (1H23), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130167PEATT594
Description
Ribonuclease Inhibitor Monoclonal specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Flow Cytometry.Specifications
Ribonuclease Inhibitor | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
RNH1 | |
Ribonuclease inhibitor 1(RNH1) (7-461aa) purified from E. coli | |
0.1 mL | |
Primary | |
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human RNH1 protein. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
1H23 | |
Flow Cytometry | |
Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054 | |
Mouse | |
Protein G purified | |
metabolism, Proteases & Other Enzymes | |
6050.0 | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction