Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribonuclease Inhibitor Antibody (OTI1B7), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273896MFV500
Description
Ribonuclease Inhibitor Monoclonal antibody specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Flow CytometrySpecifications
Ribonuclease Inhibitor | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI1B7 | |
Flow Cytometry | |
Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054 | |
Full length human recombinant protein of human RNH1 (NP_002930) produced in HEK293T cell. | |
0.1 mL | |
metabolism, Proteases & Other Enzymes | |
6050 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction