Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribonuclease Inhibitor Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299628PE
Description
Ribonuclease Inhibitor Polyclonal antibody specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Ribonuclease Inhibitor | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Ribonuclease Inhibitor. | |
0.1 mL | |
metabolism, Proteases & Other Enzymes | |
6050 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction