Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribophorin I Antibody (OTI3H3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273898PCP
Description
Ribophorin I Monoclonal specifically detects Ribophorin I in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
Ribophorin I | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
RPN1 | |
Full length recombinant protein of human Ribophorin I (NP_002941) produced in HEK293T cells. | |
0.1 mL | |
6184.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI3H3 | |
Flow Cytometry | |
DKFZp686B16177, dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit, Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1, EC 2.4.1.119, oligosaccharyltransferase 1 homolog, OST1, ribophorin IRBPH1, ribophorin-1, RPN-I | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction