Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ribophorin I Monoclonal Antibody (4B1-1E6)
GREENER_CHOICE

Catalog No. PIMA519852
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519852 Supplier Invitrogen™ Supplier No. MA519852
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: ASSEAPPLIN EDVKRTVDLS SHLAKVTAEV VLAHLGGGST SRATSFLLAL EPELEARLAH LGVQVKGEDE EENNLEVRET KIKGKSGRFF TVKLPVALDP GAKISVIVET VYTHVLHPYP TQITQSEKQF VVFEGNHYFY SPYPTKTQTM RVKLASRNVE SYTKLGNPTR SEDLLDYGPF RDVPAYSQDT FKVHYENNSP FLTITSMTRV IEVSHWGNIA VEENVDLKHT GAVLKGPFSR YDYQRQPDSG ISSIRSFKTI LPAAAQDVYY RDEIGNVSTS HLLILDDSVE MEIRPRFPLF GGWKTHYIVG YNLPSYEYLY NLGDQYALKM RFVDHVFDEQ VIDSLTVKII LPEGAKNIEI DSPYEISRAP DELHYTYLDT SGRPVIVAYK KNLVEQHIQD IVVHYTFNKV LMLQEPLLVV AAFYILFFTV IIYVRLDFSI TKDPAAEARM KVACITEQVL TLVNKRIGLY RHFDETVNRY KQSRDISTLN SGKKSLEAEH KALTSEIALL QSRLKTEGSD LCDRVSEMQK LDAQVKELVL KSAVEAERLV AGKLKKDTYI ENEKLISGKR QELVTKIDHI LDAL.

Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombinant Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribophorin I
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4B1-1E6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RPN1
Gene Accession No. P04843
Gene Alias AU018702; D6Wsu137e; Dolichyl-diphosphooligosaccharide--protein; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); OST1; QccE-21639; QtsA-18349; RBPH1; RIBI; ribophorin I; Ribophorin-1; RPN1; Rpn-1; RPN-I; unnamed protein product
Gene Symbols RPN1
Host Species Mouse
Immunogen RPN1 (AAH10839, 24 a.a. ~ 607 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6184
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.