Learn More
Invitrogen™ Ribophorin I Polyclonal Antibody

Description
Immunogen sequence: DVKRTVDLSSH LAKVTAEVVL AHLGGGSTSR ATSFLLALEP ELEARLAHLG VQVKGEDEEE NNLEVRETKI KGKSGRFFTV KLPVALDPGA KISVIVETVY THVLHPYPTQ ITQSEKQFVV FEGNHYFYSP YPTKTQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
Specifications
Specifications
| Antigen | Ribophorin I |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RPN1 |
| Gene Accession No. | P04843 |
| Gene Alias | AU018702; D6Wsu137e; Dolichyl-diphosphooligosaccharide--protein; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); OST1; QccE-21639; QtsA-18349; RBPH1; RIBI; ribophorin I; Ribophorin-1; RPN1; Rpn-1; RPN-I; unnamed protein product |
| Gene Symbols | RPN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.