Learn More
Invitrogen™ Ribophorin I Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588589
Description
Immunogen sequence: APGSASSEAP PLINEDVKRT VDLSSHLAKV TAEVVLAHLG GGSTSRATSF LLALEPELEA RLAHLGVQVK GEDEEENNLE VRETKIKGKS GRFFTVKLPV ALDPGAKISV IVETVYTHVL HPYPTQITQS EKQFVVFEGN HYFYSPYPTK TQTMRVKLAS RNVESYTKLG NPTRSEDLLD YGPFRDVPAY SQDTFKVHYE NNSPFLTITS MTRVIEVSHW GNIAVEENVD LKHTGAVLKG PFSRYDYQRQ PDSGISSIRS F; Positive Samples: HL-60, HepG2, SKOV3, MCF7, BxPC-3, Mouse liver, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Melanosome, Single-pass type I membrane protein.
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombinant Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
Specifications
Ribophorin I | |
Polyclonal | |
Unconjugated | |
RPN1 | |
AU018702; D6Wsu137e; Dolichyl-diphosphooligosaccharide--protein; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); OST1; QccE-21639; QtsA-18349; RBPH1; RIBI; ribophorin I; Ribophorin-1; RPN1; Rpn-1; RPN-I; unnamed protein product | |
Rabbit | |
Affinity Chromatography | |
RUO | |
103963, 25596, 6184 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04843, P07153, Q91YQ5 | |
RPN1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-280 of human RPN1 (NP_0029411). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.