Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribophorin II Antibody (OTI1G9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273907AF405
Description
Ribophorin II Monoclonal specifically detects Ribophorin II in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Ribophorin II | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RPN2 | |
Full length recombinant protein of human Ribophorin II (NP_002942) produced in HEK293T cells. | |
0.1 mL | |
6185.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1G9 | |
Western Blot, Immunohistochemistry | |
RIBIIREC 2.4.1.119, ribophorin IIDolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit, Ribophorin-2, RPNII, RPN-IIdolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2, SWP1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction