Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ribophorin II Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597018 Supplier Invitrogen™ Supplier No. PA597018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANTVELRVKI STEVGITNVD LSTVDKDQSI APKTTRVTYP AKAKGTFIAD SHQNFALFFQ LVDVNTGAEL TPHQTFVRLH NQKTGQEVVF VAEPDNKNVY KFELDTSERK IEFDSASGTY TLYLIIGDAT LKNPILWNVA DVVIKFPEEE APSTVLSQNL FTPKQEIQHL FREPEKRPPT V; Positive Samples: HepG2, 293T, Jurkat, MCF7, NIH/3T3, Mouse liver, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribophorin II
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPN2
Gene Accession No. P04844, P25235, Q9DBG6
Gene Alias 1300012C06Rik; AV261018; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2; hypothetical protein LOC534231; oligosaccharyltransferase complex subunit (non-catalytic); RIBIIR; ribophorin 2; ribophorin II; Ribophorin-2; RPN2; Rpn-2; RPNII; RPN-II; SWP1
Gene Symbols RPN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 360-540 of human RPN2 (NP_0029422).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20014, 6185, 64701
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.