Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ RiboRuler High Range RNA Ladder RTU PROMO
Click to view available options
Size Range:
200 to 6000 bp
Description
- Sharp bands of uniform intensity
- Easy-to-remember band sizes and quantities
- Predetermined concentration of ladder bands allows for approximate RNA quantification
- Supplied with 2X RNA Loading Dye
- Stable for 6 months at -20°C
Specifications
Specifications
Concentration | 2X |
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide |
Product Type | High Range DNA Ladder |
Size Range | 200 to 6000 bp |
Product Line | RiboRuler |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction