Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribosomal protein L26 Antibody, DyLight 680, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP222233FR
Description
Ribosomal protein L26 Polyclonal antibody specifically detects Ribosomal protein L26 in Human, Mouse samples. It is validated for Western BlotSpecifications
Ribosomal protein L26 | |
Polyclonal | |
Western Blot | |
ribosomal protein L26,60S ribosomal protein L26 | |
The immunogen this antibody was made to, maps to a region between residue 95 and 145 of human Ribosomal Protein L26 using the numbering given in entry NP_000978.1 (GeneID 6154). | |
0.1 mL | |
Cellular Markers | |
6154 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction