Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribosomal Protein S6/RPS6 Antibody (0018) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$140.50 - $393.00
Specifications
Antigen | Ribosomal Protein S6/RPS6 |
---|---|
Clone | 0018 |
Dilution | Western Blot 1:1000-1:5000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western BlotSpecifications
Ribosomal Protein S6/RPS6 | |
Western Blot 1:1000-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
The antibody is produced by immunizing rabbits with a synthetic phosphopeptide corresponding to residues around Ser240, Ser244) of Human phospho-Ribosomal Protein S6/RPS6 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0018 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control | |
0.2 um filtered solution in PBS | |
6194 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title