Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribosomal Protein S6/RPS6 Antibody (522731), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5436M
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
Ribosomal Protein S6/RPS6 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
522731 | |
Western Blot, Immunocytochemistry | |
Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
E. coli-derived recombinant human RPS6, Met1-Lys249, Accession # P62753 | |
0.1 mL | |
Cancer, DNA Repair, mTOR Pathway, Translation Control | |
6194 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction