Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ribosome-inactivating protein Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598352
Description
Ribosome-inactivating protein Polyclonal Antibody for Western Blot, ELISA
Has cytotoxic activity towards cancer cells, but not normal cells. Inhibits the growth of the human leukemia cell line K562, the murine melanoma cell line B16 and the lung adenocarcinoma cell line A549 with IC50 values of 88.1 nM, 63.4 nM and 359.3 nM respectively. [Uniprot].
Specifications
Ribosome-inactivating protein | |
Polyclonal | |
Unconjugated | |
cucurmosin; Ribosome-inactivating protein cucurmosin; rRNA N-glycosidase | |
Recombinant Cucurbita moschata Ribosome-inactivating protein cucurmosin protein (1-244AA). | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Plant | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction