Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribosome maturation protein SBDS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Rabbit |
Description
Ribosome maturation protein SBDS Polyclonal specifically detects Ribosome maturation protein SBDS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Q9Y3A5 | |
51119 | |
Synthetic peptide directed towards the N terminal of human SBDS (NP_057122). Peptide sequence LQTHSVFVNVSKGQVAKKEDLISAFGTDDQTEICKQILTKGEVQVSDKER. | |
Primary | |
28 kDa |
Polyclonal | |
Purified | |
RUO | |
FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS | |
SBDS | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title