Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ribulose-phosphate 3-epimerase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544605 Supplier Invitrogen™ Supplier No. PA544605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MMVSKPEQWV KPMAVAGANQ YTFHLEATEN PGALIKDIRE NGMKVGLAIK Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Catalyzes the reversible epimerization of D-ribulose 5-phosphate to D-xylulose 5-phosphate. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribulose-phosphate 3-epimerase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RPE
Gene Accession No. Q96AT9
Gene Alias 2810429B02Rik; 5730518J08Rik; HUSSY-17; ik:tdsubc_2c8; PE; Ribulose-5-phosphate-3-epimerase; ribulose-5-phosphate-epimerase; Ribulose-phosphate 3-epimerase; Rpe; R-PE; RPE2-1; R-phycoerythrin; wu:fa07h08; wu:fb93d11; wu:fc20b11; xx:tdsubc_2c8
Gene Symbols RPE
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RPE.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6120
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.