Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ribulose-phosphate 3-epimerase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530153
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (96%), Zebrafish (84%), Xenopus laevis (90%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalyzes the reversible epimerization of D-ribulose 5-phosphate to D-xylulose 5-phosphate. [UniProt].
Specifications
Ribulose-phosphate 3-epimerase | |
Polyclonal | |
Unconjugated | |
RPE | |
2810429B02Rik; 5730518J08Rik; HUSSY-17; ik:tdsubc_2c8; PE; Ribulose-5-phosphate-3-epimerase; ribulose-5-phosphate-epimerase; Ribulose-phosphate 3-epimerase; Rpe; R-PE; RPE2-1; R-phycoerythrin; wu:fa07h08; wu:fb93d11; wu:fc20b11; xx:tdsubc_2c8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6120 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M Tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96AT9 | |
RPE | |
Recombinant fragment corresponding to a region within amino acids 42 and 228 of Human RPE. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction