Learn More
Ric-8A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304571AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins. Able to activate GNAI1, GNAO1 and GNAQ, but not GNAS by exchanging bound GDP for free GTP. Involved in regulation of microtubule pulling forces during mitotic movement of chromosomes by stimulating G(i)-alpha protein, possibly leading to release G(i)-alpha-GTP and NuMA proteins from the NuMA-GPSM2-G(i)-alpha-GDP complex. Also acts as an activator for G(q)-alpha (GNAQ) protein by enhancing the G(q)-coupled receptor-mediated ERK activation.Specifications
Ric-8A | |
Polyclonal | |
Unconjugated | |
RIC8A | |
RIC8A; synembryn; resistance to inhibitors of cholinesterase 8 homolog A (C. elegans); MGC104517; MGC131931; RIC8; RIC8 protein; resistance to inhibitors of cholinesterase 8 homolog A; MGC148073; MGC148074 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60626 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9NPQ8 | |
RIC8A | |
Between 1 and 50 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.