Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIC8A Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB272091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272091 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272091 Supplier Novus Biologicals Supplier No. NBP236536AF350
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIC8A Polyclonal antibody specifically detects RIC8A in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen RIC8A
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A
Host Species Rabbit
Immunogen The immunogen recognized by the antibody maps to a region between residue 418 to 468 of human Resistance to Inhibitors of Cholinesterase 8 Homolog A using the numbering given in entry EAW61245.1.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60626
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.