Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIC8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110295
Description
Immunogen sequence: DVRTRPEVGE MLRNKLVRLM THLDTDVKRV AAEFLFVLCS ESVPRFIKYT GYGNAAGLLA ARGLMAGGRP EGQYSEDEDT DTDEYKEAKA SINPVTGRVE EKPPNPMEGM TEEQKEHEAM KLVTMFDKLS RNRVIQPMGM SPRGHLTSLQ DAMCETMEQQ LSSDPDSDPD.
Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins. Able to activate GNAI1, GNAO1 and GNAQ, but not GNAS by exchanging bound GDP for free GTP. Involved in regulation of microtubule pulling forces during mitotic movement of chromosomes by stimulating G(i)-alpha protein, possibly leading to release G(i)-alpha-GTP and NuMA proteins from the NuMA-GPSM2-G(i)-alpha-GDP complex. Also acts as an activator for G(q)-alpha (GNAQ) protein by enhancing the G(q)-coupled receptor-mediated ERK activation.
Specifications
| RIC8A | |
| Polyclonal | |
| Unconjugated | |
| RIC8A | |
| AI114950; heterotrimeric G protein guanine nucleotide exchange factor Ric-8A; MGC104517; MGC131931; MGC148073; MGC148074; Protein Ric-8A; resistance to inhibitors of cholinesterase 8 homolog; resistance to inhibitors of cholinesterase 8 homolog (C. elegans); resistance to inhibitors of cholinesterase 8 homolog A; resistance to inhibitors of cholinesterase 8A; Ric8; RIC-8; RIC8 guanine nucleotide exchange factor A; RIC8 protein; Ric8a; Ric-8A; synembryn; synembryn-A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 101489, 293614, 60626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TIR3, Q80ZG1, Q9NPQ8 | |
| RIC8A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 362-531 of human RIC8A (NP_001273063.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction