Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIC8A Rabbit anti-Human, Mouse, Polyclonal, FabGennix

Catalog No. 502208849
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8849 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8849 Supplier Fabgennix Inc Supplier No. RIC8A101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIC8A Polyclonal antibody specifically detects RIC8A in Human, Mouse samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen RIC8A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene RIC8A
Gene Accession No. Q3TIR3, Q9NPQ8
Gene Alias AI114950; heterotrimeric G protein guanine nucleotide exchange factor Ric-8A; MGC104517; MGC131931; MGC148073; MGC148074; Protein Ric-8A; resistance to inhibitors of cholinesterase 8 homolog; resistance to inhibitors of cholinesterase 8 homolog (C. elegans); resistance to inhibitors of cholinesterase 8 homolog A; resistance to inhibitors of cholinesterase 8A; Ric8; RIC-8; RIC8 guanine nucleotide exchange factor A; RIC8 protein; Ric8a; Ric-8A; synembryn; synembryn-A
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acids 500-535 on RIC8A protein
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101489, 60626
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.