Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIC8B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | RIC8B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RIC8B Polyclonal specifically detects RIC8B in Human samples. It is validated for Western Blot.Specifications
RIC8B | |
Polyclonal | |
Rabbit | |
A2RTZ0 | |
55188 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Brain synembrin, hSyn, Protein Ric-8B, RIC8, RIC8B resistance to inhibitors of cholinesterase 8 homolog B (C. elegans), synembryn, synembryn-B | |
RIC8B | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title