Learn More
Invitrogen™ RIC8B Polyclonal Antibody

Description
RIC8B Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RIC8B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RIC8B |
| Gene Accession No. | Q6DRJ9, Q9NVN3 |
| Gene Alias | BC051080; brain synembrin; brain synembryn; fj97c10; heterotrimeric G protein guanine nucleotide exchange factor-like protein Ric-8B; hSyn; Protein Ric-8B; resistance to inhibitors of cholinesterase 8 homolog B; resistance to inhibitors of cholinesterase 8 homolog B (C. elegans); resistance to inhibitors of cholinesterase 8B; RIC8; Ric-8; RIC8 guanine nucleotide exchange factor B; ric8b; Ric-8b; synbl; synembryn like; synembryn-B; Synembryn-like; Unknown (protein for IMAGE:7408979); wu:fj97c10 |
| Gene Symbols | RIC8B |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.