Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIC8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104334 Supplier Invitrogen™ Supplier No. PA5104334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RIC8B.

Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins by exchanging bound GDP for free GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIC8B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RIC8B
Gene Accession No. Q80XE1, Q80ZG0, Q9NVN3
Gene Alias BC051080; brain synembrin; brain synembryn; fj97c10; heterotrimeric G protein guanine nucleotide exchange factor-like protein Ric-8B; hSyn; Protein Ric-8B; resistance to inhibitors of cholinesterase 8 homolog B; resistance to inhibitors of cholinesterase 8 homolog B (C. elegans); resistance to inhibitors of cholinesterase 8B; RIC8; Ric-8; RIC8 guanine nucleotide exchange factor B; ric8b; Ric-8b; synbl; synembryn like; synembryn-B; Synembryn-like; Unknown (protein for IMAGE:7408979); wu:fj97c10
Gene Symbols RIC8B
Host Species Rabbit
Immunogen A synthesized peptide derived from human RIC8B(Accession Q9NVN3), corresponding to amino acid residues D464-E514.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237422, 314681, 55188
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.