Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cleaning Solution, Chromic-Sulfuric Acid Solution, Ricca Chemical
SDP

Catalog No. 215032 Shop All Ricca Chemical Company Products
Click to view available options
Volume (Metric):
500 mL
1 L
2.5 L

For Cleaning Laboratory Glassware

  • Clear, brown liquid
  • 7789-12-0 (Sodium Dichromate Dihydrate); 1333-82-0 (Chromium (VI) Trioxide); 7664-93-9 (Sulfuric Acid)

Specifications

Product Type Cleaning Solution
Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Appearance Dark Brown
Boiling Point ∼300°C
Certifications/Compliance OSHA, 29 CFR 1910.1200, TSCA
Composition Chromium (VI) Trioxide 0.8 to 1%, Sulfuric Acid 97 to 99 %, Water, Deionized 1 to 1.2%, Sodium Dichr
Content And Storage 15°C to 30°C
Volume (Metric) 1 L
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.