Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate, Copper Sulfate, 1.055 Specific Gravity at 25°C (77°F), Ricca Chemical
SDP

Catalog No. p-3247787 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2340-16 500 mL
2340-32 1 L
2340-1 4 L
2340-5 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2340-16 Supplier Ricca Chemical Company Supplier No. 234016
Only null left
Add to Cart
Add to Cart

For Hemoglobin Estimation in Men

  • 1.055 Specific Gravity at 25°C (77°F)
  • CAS: 7758-99-8 (Copper Sulfate Pentahydrate); 7732-18-5 (Water, Deionized)

Screening of male blood donors

Specifications

Content And Storage 15°C to 30°C
CE Marker No
Type Copper Sulfate
For Use With (Application) Screening of male Blood donors
Packaging Type Amber Glass
pH 4
Quantity 500 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.