Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Modified Davidson's Fixative II, Ricca Chemical

Click to view available options
Quantity:
500 mL
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
Specifications
Specifications
Physical Form | Solution |
Packaging | Natural Poly Bottle |
Color | Blue |
pH | 2.41 to 2.54 |
Quantity | 500 mL |
Grade | Laboratory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction