Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RICH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558550 Supplier Invitrogen™ Supplier No. PA558550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STPSPYGLSY PQGYSLASGQ LSPAAAPPLA SPSVFTSTLS KSRPTPKPRQ RPTLPPPQPP TVNLSASSPQ STEAPMLDGM SPGESMSTDL VHFDIPSIHI ELGSTLRLSP LEHMRRHSVT DKRDSE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

GTPase-activating protein (GAP) that stimulates the GTPase activity of Rho-type GTPases. Thereby, controls Rho-type GTPases cycling between their active GTP-bound and inactive GDP-bound states. Acts as a GAP at least for CDC42 and RAC1 (PubMed:11431473). In neurons, is involved in dendritic spine formation and synaptic plasticity in a specific RAC1-GAP activity. Limits the initiation of exploratory dendritic filopodia. Recruited to actin-patches that seed filopodia, binds specifically to plasma membrane sections that are deformed inward by acto-myosin mediated contractile forces. Acts through GAP activity on RAC1 to reduce actin polymerization necessary for filopodia formation. In association with SHANK3, promotes GRIA1 exocytosis from recycling endosomes and spine morphological changes associated to long-term potentiation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RICH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP44
Gene Accession No. Q17R89
Gene Alias ARHGAP44; KIAA0672; NPC-A-10; Rho GTPase activating protein 44; rho GTPase-activating protein 44; rho GTPase-activating protein RICH2; RhoGAP interacting with CIP4 homologs protein 2; Rho-type GTPase-activating protein RICH2; RICH2; RICH-2
Gene Symbols ARHGAP44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RICH2. Recombinant protein control fragment (Product #RP-102315).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9912
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.