Learn More
Epredia™ Formaldehyde Warning Labels For Specimen Containers
Catalog No. 22046310 Shop All Epredia Products
Click to view available options
Closure Included:
Yes
This item is not returnable. View return policy
This item is not returnable. View return policy
Two sizes of formaldehyde warning labels to complete any specimen collection needs.
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
  • Easy to transport trays ideal for operating rooms and hospitals
  • Clear polystyrene containers have screw-top lids
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
  • Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Easy to transport trays
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
  • Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Stack pack reduces excess packaging
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Product Type Specimen Container Label
Closure Included Yes
Description Formalin Warning Labels, Small
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.