Learn More
Description
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
Specifications
Specifications
| Packaging Type | 4 oz Dispenser Bottle |
| Product Line | Cytoseal XYL |
| Product Type | Mountant |
| Quantity | 4 oz. |
| Formulation | Fast-drying formula reduces waiting time and increases throughput |
| For Use With (Application) | Provides exceptional clarity and longevity for your microscopy slides |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.