Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Dehydrant Alcohol
Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
1.32 gal.
55 gal. drum
Product Type:
Alcohol
Formulation:
100% Dehydrant
70% Dehydrant
80% Dehydrant
95% Dehydrant
Description
- Blend of ethyl, methyl and isopropyl alcohols
- Can be used as a dye or stain solvent
- Miscible with all clearing agents
Specifications
Specifications
| Product Type | Alcohol |
| Quantity | 1 gal. |
| Formulation | 70% Dehydrant |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction