Learn More
Epredia™ Richard-Allan Scientific™ Gram Stain (Tissue) Kit, Crystal Violet Solution
Click to view available options
Type:
Crystal Violet
Gram Stain (Tissue) Kit
Safranin O Stain Solution
Description
- Gram-positive organisms stain blue to blue-black
- Gram-negative organisms stain red
- Tartrazine provides a yellow background stain
Specifications
Specifications
Type | Crystal Violet |
Quantity | 500mL |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov